missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc ICP Standard, 1mL = 1mg Zn (1,000ppm Zn), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPZN1KN100

 View more versions of this product

Catalog No. PZN1KN-100



Specifications

Specifications

7440-66-6,7697-37-2,7732-18-5
97.0,4.81,0.1
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Liquid
1,000ppm Zn
Colorless
93.0,4.63,0.09
Zn
[Zn]
65.38
CHEBI:30185
Poly natural
1 mL = 1 mg Zn
100mL

Chemical Identifiers

7440-66-6
65.38
23994
zinc
Zn
HCHKCACWOHOZIP-UHFFFAOYSA-N
CHEBI:30185
[Zn]
Documents