missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc ICP Standard, 1mL = 1mg Zn (1,000ppm Zn), Ricca Chemical

$180.33 - $333.85

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PZN1KN-100
View Documents
Ricca Chemical Company
RIPZN1KN100
100mL Poly natural Each for $180.33
 
PZN1KN-500
SDS View Documents
Ricca Chemical Company
RIPZN1KN500
500mL Poly natural Each for $333.85
 
Specifications

Chemical Identifiers

7440-66-6
65.38
23994
zinc
Zn
HCHKCACWOHOZIP-UHFFFAOYSA-N
CHEBI:30185
[Zn]

Specifications

7440-66-6,7697-37-2,7732-18-5
97.0,4.81,0.1
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Liquid
1,000ppm Zn
Colorless
Zinc ICP Standard
93.0,4.63,0.09
Zn
[Zn]
65.38
CHEBI:30185
Poly natural
1 mL = 1 mg Zn
100mL