Please login to your online account to display your discounted pricing

Zinc, Granular, 30 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  424801

 View more versions of this product

Catalog No. 02004599

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 65.39
Purity Grade Baker Analyzed™ ACS Reagent
Mesh Size 30 Mesh
Chemical Name or Material Zinc
Physical Form Granular
Molecular Formula Zn
CAS 7440-66-6
Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 7.14kg

Specifications

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Zn
Formula Weight 65.39