missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc, granular, 30 mesh, ACROS Organics™

$47.16 - $130.81

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222610025 SDS View Documents Acros Organics
AC222610025
2.5kg Plastic bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC222611000 SDS View Documents Acros Organics
AC222611000
100g Plastic bottle Each for $47.16
 
AC222615000 SDS View Documents Acros Organics
222615000
500g Plastic bottle Each for $130.81
 
Description

Description

Reduction reagent
Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

2.5kg
0.1ppm max.
Zn
MFCD00011291
100ppm max.
15, 10326
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
Plastic bottle
CHEBI:30185
Fine Granular Powder
Gray
907.0°C
7440-66-6
7.1400g/mL
Zn
100ppm max.
01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
23994
65.38
≥99.7%
420.0°C
Zinc, 30 mesh, p.a.
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-175-3

RTECSNumber : ZG8600000

TSCA : TSCA