missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc, granular, 30 mesh, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 222615000

 View more versions of this product

Catalog No. AC222615000


 

Specifications Description & Specifications

Specifications

Zinc
30 mesh, p.a.
0.1ppm max.
Fine Granular Powder
100ppm max.
100ppm max.
65.38
01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Plastic bottle
500g
7440-66-6
7.1400g/mL
≥99.7%
Zn
Zn
MFCD00011291
15, 10326
Gray

Chemical Identifiers

7440-66-6
Zn
MFCD00011291
≥99.7%
Zn
65.38
Zinc, 30 mesh, p.a.

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-175-3

RTECSNumber : ZG8600000

TSCA : TSCA