Please login to your online account to display your discounted pricing

Zinc, granular, 30 mesh, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC222611000

 View more versions of this product

Catalog No. AC222611000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.7%
Physical Form Fine Granular Powder
Formula Weight 65.38
Quantity 100g
Color Gray
Merck Index 15, 10326
EINECS Number 231-175-3
TSCA TSCA
Density 7.1400g/mL
RTECS Number ZG8600000
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Chemical Name or Material Zinc
Iron (Fe) 100ppm max.
Linear Formula Zn
CAS 7440-66-6
Arsenic (As) 0.1ppm max.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 30 mesh, p.a.
Molecular Formula Zn
Lead (Pb) 100ppm max.
MDL Number MFCD00011291
Packaging Plastic bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material Zinc
Name Note 30 mesh, p.a.
CAS 7440-66-6
Assay ≥99.7%
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.38
MDL Number MFCD00011291
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents