Please login to your online account to display your discounted pricing

Zinc (Granular, 20 Mesh/Certified ACS), Fisher Chemical


Low in arsenic, iron and lead

Quantity 3kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  Z153

 View more versions of this product

Catalog No. Z15-3

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Boiling Point 907°C
Melting Point 419°C
Purity Grade Certified ACS Grade
Formula Weight 65.41
Quantity 3kg
Mesh Size 20 Mesh
Physical Form Solid
Vapor Pressure 1mmHg at 487°C
Color Light Blue
Density 7.14g/cm³
Atomic Weight 65.37
Assay Percent Range ≥99.8 %
Identification Pass Test
Molecular Formula Zn
Chemical Name or Material Zinc
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
CAS 7440-66-6
MDL Number MFCD00011291
Packaging Poly Bottle
Lead (Pb) 0.01% max.
Iron (Fe) 0.01% max.
Additional Information Vapor Pressure: 1mmHg at 487°C
Suitability Pass Test
Health Hazard 2 CAUTION!
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥99.8%
Assay Percent Range ≥99.8 %
Molecular Formula Zn
Formula Weight 65.41
MDL Number MFCD00011291
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents