Please login to your online account to display your discounted pricing

Zinc (Granular, 20 Mesh/Certified ACS), Fisher Chemical

Low in arsenic, iron and lead

$420.77 - $1,713.92

Chemical Identifiers

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥99.8%
Assay Percent Range ≥99.8 %
Molecular Formula Zn
Formula Weight 65.41
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

Z15-3

  SDS
fisher scientific company
Z153
Poly Bottle 3kg Each for $1,713.92
Case of 6 Each for $10,283.52

Z15-500

  SDS
fisher scientific company
Z15500
Glass Bottle 500g Each for $420.77
Case of 6 Each for $2,524.62
Description & Specifications

Specifications

Assay ≥99.8%
Purity Grade Certified ACS Grade
Formula Weight 65.41
Melting Point 419°C
Boiling Point 907°C
Quantity 3kg
Density 7.14g/cm³
Physical Form Solid
Mesh Size 20 Mesh
Vapor Pressure 1mmHg at 487°C
Atomic Weight 65.37
Color Light Blue
Chemical Name or Material Zinc
Identification Pass Test
Assay Percent Range ≥99.8 %
Molecular Formula Zn
CAS 7440-66-6
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Additional Information Vapor Pressure: 1mmHg at 487°C
Iron (Fe) 0.01% max.
Lead (Pb) 0.01% max.
MDL Number MFCD00011291
Packaging Poly Bottle
Health Hazard 2 CAUTION!
Suitability Pass Test
ChemAlert Storage Symbol Gray

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!