Please login to your online account to display your discounted pricing

Zinc, granular, 20 mesh, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  222601000

 View more versions of this product

Catalog No. AC222601000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.7% (Complexometry)
Formula Weight 65.38
Physical Form Granular Powder
Quantity 100g
Color Gray
Density 7.1400g/mL
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Arsenic (As) 0.1ppm max.
Chemical Name or Material Zinc
CAS 7440-66-6
Lead (Pb) 100ppm max.
Packaging Plastic bottle
Iron (Fe) 100ppm max.
Linear Formula Zn
Molecular Formula Zn

Specifications

Chemical Name or Material Zinc
Assay ≥99.7% (Complexometry)
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.38