missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2302222

 View more versions of this product

Catalog No. AAB2302222



Description

Description

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Zinc gluconate hydrate
6.5
C12H22O14Zn
100 g
zinc ii gluconate hydrate, zinc 2+ bis d-gluconate hydrate
WHMDKBIGKVEYHS-IYEMJOQQSA-L
zinc;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate
131674739
97%
4468-02-4
Odorless
MFCD00868110
14,4456
Soluble in water 100g/ l.
[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
455.67
455.69 (Anhydrous)

Chemical Identifiers

4468-02-4
MFCD00868110
Safety and Handling

Safety and Handling

EINECSNumber : 224-736-9

RTECSNumber : ZH3750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only