missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc foil, 99.994% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 4.5g/50 x 50mm

Manufacturer:  Alfa Aesar 11912HG

 View more versions of this product

Catalog No. AA11912HG



Specifications

Specifications

Zinc
7440-66-6
99.994% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Gray
100 x 300mm
Gray foil
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
Odorless
23994
Foil, 0.25mm (0.01 in.) thick

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

Documents