missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc foil, 99.95% (metals basis), Alfa Aesar™

Form: approx. 53g/30x10cm

Manufacturer:  Thermo Fisher Scientific 10436CB

 View more versions of this product

Catalog No. AA10436CB



Specifications

Specifications

Zinc
7440-66-6
99.98% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Gray
250cm
Gray foil
Zn
blue powder
[Zn]
Odorless
23994
Foil, 0.25mm (0.01 in.) thick, 30cm (12 in.) wide

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
blue powder
CHEBI:30185
[Zn]
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

Documents