missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Foil, 1.6mm (0.063 in.) thick, 15 x 15cm (5.9 x 5.9 in.), 99% (Metals basis), Alfa Aesar™

Form: Approx.255g/15 x 15cm

Manufacturer:  Thermo Fisher Scientific 10438KG

 View more versions of this product

Catalog No. AA10438KG



Specifications

Specifications

Zinc
7440-66-6
7.14g/cm3
Zn
blue powder
[Zn]
Odorless
23994
Foil, 1.6mm (0.063 in.) thick, 15 X 15cm (5.9 X 5.9 in.)
1 pc.
Gray foil
99% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Gray

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
blue powder
CHEBI:30185
[Zn]
Documents