missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.9985% (Metals basis), Alfa Aesar™

Form: Approx.4.5g/25 x 25mm

Manufacturer:  Alfa Aesar 11915GH

 View more versions of this product

Catalog No. AA11915GH



Specifications

Specifications

Zinc
7440-66-6
99.9985% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Gray
100 x 100mm
Gray foil
Zn
blue powder
[Zn]
Odorless
23994
Foil, 1.0mm (0.04 in.) thick

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
blue powder
CHEBI:30185
[Zn]
Documents