missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Foil, 0.62mm (0.024 in.) thick, 30cm (12 in.) wide, 99.9% (Metals basis), Alfa Aesar™

Form: Approx.665g/30 x 50cm

Manufacturer:  Alfa Aesar 10437BW

 View more versions of this product

Catalog No. AA10437BW



Specifications

Specifications

Zinc
7440-66-6
99.9% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
CHEBI:30185
Gray
50cm
Gray foil
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
Odorless
23994
Foil, 0.62mm (0.024 in.) thick, 30cm (12 in.) wide

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Documents