missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc, Dust Particle Size <63μm, MilliporeSigma™

Manufacturer:  MilliporeSigma 1087749050

 View more versions of this product

Catalog No. M1087749050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Zinc
7440-66-6
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
Odorless
Steel drum
CHEBI:30185
Solid
50kg
Gray solid
UN 3077
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
65.38
23994
65.39
1.33hPa

Chemical Identifiers

7440-66-6
65.38
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Zn
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Safety and Handling

Safety and Handling

H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-175-3

Documents