missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc diethyldithiocarbamate, Zn 17-19.5%

Chelating agent

Manufacturer:  Alfa Aesar™ 18697A3

 View more versions of this product

Catalog No. AA18697A3



Specifications

Specifications

Zinc diethyldithiocarbamate
1.48g/mL
Zn[CS2N(CH2CH3)2]2
MFCD00064798
UN3077
2kg
Zn 17 to 19.5%
Diethyldithiocarbamic acid zinc salt; Zinc bis(diethyldithiocarbamate)
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]
361.904
361.9
14324-55-1
C10H20N2S4Zn
178°C to 183°C
White
7.8
Odorless
204° (400°F)
RKQOSDAEEGPRER-UHFFFAOYSA-L
zinc;N,N-diethylcarbamodithioate
26633
Crystalline

Chemical Identifiers

14324-55-1
361.904
RKQOSDAEEGPRER-UHFFFAOYSA-L
26633
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]
C10H20N2S4Zn
MFCD00064798
Diethyldithiocarbamic acid zinc salt; Zinc bis(diethyldithiocarbamate)
zinc;N,N-diethylcarbamodithioate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P280g-P262-P273-P501a
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ZH0350000

TSCA : TSCA

SDS Documents