Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc cyclohexanebutyrate dihydrate, AAS

Chemical Identifiers

Chemical Name or Material Zinc cyclohexanebutyrate dihydrate
CAS 38582-18-2
Name Note AAS
Formula Weight 439.91 (403.87 Anhydrous)
MDL Number MFCD00035537
Synonym Cyclohexanebutyric acid zinc salt dihydrate; 4-Cyclohexylbutyric acid zinc salt dihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3973403

 
alfa aesar™
3973403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3973406

 
alfa aesar™
3973406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 439.91 (403.87 Anhydrous)
Chemical Name or Material Zinc cyclohexanebutyrate dihydrate
TSCA TSCA
Name Note AAS
Melting Point 77° to 80°C
Physical Form Powder
MDL Number MFCD00035537
CAS 38582-18-2
Synonym Cyclohexanebutyric acid zinc salt dihydrate; 4-Cyclohexylbutyric acid zinc salt dihydrate
Quantity 1g