Learn More
Zinc cyanide, 98%, pure
CAS: 557-21-1 | C2N2Zn | 117.43 g/mol
$121.64 - $317.73
Chemical Identifiers
| CAS | 557-21-1 |
|---|---|
| Molecular Formula | C2N2Zn |
| Molecular Weight (g/mol) | 117.43 |
| MDL Number | MFCD00011292 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC368072500
|
Thermo Scientific Chemicals
368072500 |
250 g | Plastic bottle |
Each for $121.64
|
|
||||
|
AC368070010
|
Thermo Scientific Chemicals
368070010 |
1 kg | Plastic bottle |
Each for $317.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
| 557-21-1 | |
| C2N2Zn | |
| 117.43 | |
| MFCD00011292 |
Specifications
| 800.0°C | |
| 557-21-1 | |
| 250 g | |
| C2N2Zn | |
| MFCD00011292 | |
| 15,10334 | |
| 117.43 | |
| 98% | |
| Plastic bottle |
| White | |
| Powder | |
| 97.5% min. (Complexometry) | |
| Zn(CN)2 | |
| 02,III,86 | |
| Solubility in water: insoluble | |
| 117.43 | |
| Pure | |
| Zinc cyanide, 98% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Contact with acids liberates very toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-162-9
RUO – Research Use Only