Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc cyanide, 85%

Electroplating, reagent

Chemical Identifiers

Chemical Name or Material Zinc cyanide
CAS 557-21-1
Formula Weight 117.42
MDL Number MFCD00011292
UN Number UN1713
Beilstein 4124366
Merck Index 14,10135
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1415822

  SDS
alfa aesar™
1415822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1415836

  SDS
alfa aesar™
1415836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1415857

  SDS
alfa aesar™
1415857
4 x 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 117.42
TSCA TSCA
Chemical Name or Material Zinc cyanide
UN Number UN1713
Quantity 100g
MDL Number MFCD00011292
Beilstein 4124366
Sensitivity Hygroscopic
Melting Point 800°C (dec.)
Color White
RTECS Number ZH1575000
Solubility Information Insoluble in water, alcohol; Soluble in solutions of alkali cyanides or hydroxides
Physical Form Crystalline
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Density 1.852g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ZINC CYANIDE
Merck Index 14,10135
Assay 85%
CAS 557-21-1
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.