missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, ultra dry, 99.999% (metals basis), Alfa Aesar™

$71.26 - $626.09

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc butter
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3572606 View Documents Alfa Aesar
3572606
5g Each for $71.26
 
AA3572614 View Documents Alfa Aesar
3572614
25g Each for $214.35
 
AA3572622 View Documents Alfa Aesar
3572622
100g Each for $626.09
 
Specifications

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc butter
CHEBI:49976
Cl[Zn]Cl

Specifications

7646-85-7
Cl2Zn
zinc butter
Cl[Zn]Cl
136.28
CHEBI:49976
Powder, Ampouled Under Argon
5g
99.999% (metals basis)
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.28
2.91g/mL
Zinc chloride
Safety and Handling

Safety and Handling

EINECSNumber : 231-592-0