missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc chloride, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4419814

 View more versions of this product

Catalog No. AA4419814



Description

Description

  • Ampouled under argon
Specifications

Specifications

Zinc chloride
7646-85-7
Cl2Zn
UN2331
zinc butter
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
99.999%
−10 Mesh
Ampule
732°C
25g
Ultra dry
(metals basis)
MFCD00011295
14,10132
Freely soluble in water and dilute hydrochloric acid. Soluble in alcohol,glycerol,ether and acetone.
Cl[Zn]Cl
136.28
CHEBI:49976
-10 Mesh Beads,Ampouled under argon
2.91 g/mL
Odorless
Hygroscopic
290°C

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc butter
CHEBI:49976
Cl[Zn]Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ZINC CHLORIDE, ANHYDROUS

EINECSNumber : 231-592-0

RTECSNumber : ZH1400000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents