missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, ultra dry, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1369409

 View more versions of this product

Catalog No. AA1369409



Specifications

Specifications

ultra dry
99.99% metal basis
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
Odorless
White
7646-85-7
Cl2Zn
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Cl[Zn]Cl
136.28
CHEBI:49976
Powder/Crystalline/Beads
1hPa (1mm Hg) at 20°C
10g

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
CHEBI:49976
Cl[Zn]Cl
Documents