Please login to your online account to display your discounted pricing

Zinc Chloride (Granular/USP/EP/BP/JP), Fisher Chemical

$4,563.62 - $4,563.62

Chemical Identifiers

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade BP/EP/JP/USP
Physical Form Solid
Assay ≥97.0% (JP)
Molecular Formula Cl2Zn
Formula Weight 136.3
MDL Number MFCD00011295
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

Z3412

  SDS
fisher scientific company
Z3412
Poly Pail 12kg Each for $4,563.62
Description & Specifications

Specifications

Assay ≥97.0% (JP)
Purity Grade BP/EP/JP/USP
Formula Weight 136.3
Boiling Point 732°C
Melting Point 293°C
Quantity 12kg
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula Cl2Zn
Vapor Pressure 1.3mbar at 428°C
Color White
pH 4.6 to 5.5 (EP)
Identification Pass Test (BP, EP, JP
Alkalis and Alkaline Earths ≤1.0% (JP, USP)
Oxychlorides Pass Test (BP, EP, JP
Residual Solvents Meets Requirements (BP)
Ammonium ≤400ppm (BP, EP), Pass Test (JP)
Sulfate ≤200ppm (BP, EP), ≤0.010% (JP), ≤0.03% (USP)
Bacterial Endotoxins Report EU/mg (EP)
Al Ca Heavy Metals Fe and Mg Pass Test (BP)
Endotoxin Report EU/mg (USP)
Chemical Name or Material Zinc Chloride
Lead (Pb) 0.005% max. (USP)
MDL Number MFCD00011295
CAS 7646-85-7
Arsenic (As) ≤5ppm (JP)
Heavy Metals ≤50ppm (JP)
Packaging Poly Pail
Ammonium Salts Pass Test (USP)
Additional Information Vapor Pressure: 1.3mbar at 428°C
Health Hazard 2 DANGER!
Clarity and Color Clear and colorless (JP)
ChemAlert Storage Symbol White
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:1
Instability:1
Safety and Handling
 

Emergency Overview
Causes burns by all exposure routes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:1
Instability:1

DANGER!