Please login to your online account to display your discounted pricing

Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T.Baker™

$353.20 - $1,350.27

Chemical Identifiers

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Name Note Endotoxin Tested
Purity Grade Multi-Compendial/USP
Molecular Formula ZnCl2
Formula Weight 136.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004595

 
avantor performance
432601
Poly Bottle 500g Each for $353.20

02004594

 
avantor performance
432605
Poly Bottle 2.5kg Each for $1,350.27
Description & Specifications

Specifications

Purity Grade Multi-Compendial/USP
Formula Weight 136.3
Physical Form Granular
Chemical Name or Material Zinc Chloride
Specific Gravity 1L = 2.91kg
Molecular Formula ZnCl2
Name Note Endotoxin Tested
Packaging Poly Bottle
CAS 7646-85-7
Quantity 500g