missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, anhydrous, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8790022

 View more versions of this product

Catalog No. AA8790022



Specifications

Specifications

Zinc chloride, anhydrous
99.99% (metals basis)
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.28
2.91 g/mL
7646-85-7
Cl2Zn
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Cl[Zn]Cl
136.28
CHEBI:49976
Lumps
100g

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
CHEBI:49976
Cl[Zn]Cl
Safety and Handling

Safety and Handling

EINECSNumber : 231-592-0

SDS Documents