missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, anhydrous, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8790088

 View more versions of this product

Catalog No. AA8790088



Specifications

Specifications

Zinc chloride
7646-85-7
(metals basis)
ZnCl2
UN2331
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
2.91
Hygroscopic
White
2.5g
anhydrous
99.95%
Cl2Zn
MFCD00011295
14,10132
Cl[Zn]Cl
136.28
CHEBI:49976
Lumps
Odorless
732°C
290°C

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
CHEBI:49976
Cl[Zn]Cl
Cl2Zn
MFCD00011295
5727
dichlorozinc
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302;
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ZINC CHLORIDE, ANHYDROUS

RTECSNumber : ZH1400000

TSCA : TSCA

SDS Documents