Please login to your online account to display your discounted pricing

Zinc Chloride ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  878002

 View more versions of this product

Catalog No. MK8780125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS
Physical Form Granular
Molecular Formula ZnCl2
Formula Weight 136.3g/mol

Specifications

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS
Physical Form Granular
Molecular Formula ZnCl2
Formula Weight 136.3g/mol