Please login to your online account to display your discounted pricing

Zinc Chloride ACS AR Granular, Macron Fine Chemicals™

macron

$619.40 - $619.40

Chemical Identifiers

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula ZnCl2
Formula Weight 136.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK8780212

 
avantor performance
878006
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK8780500

 
avantor performance
878004*
POLYSTORMOR 500g Each for $619.40
Case of 12 Each for $7,432.80

MK8780125

 
avantor performance
878002
POLYSTORMOR 125g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 136.3
Quantity 2.5kg
Chemical Name or Material Zinc Chloride
Packaging Poly
Physical Form Granular
Molecular Formula ZnCl2
CAS 7646-85-7