missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, ACS, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1230730

 View more versions of this product

Catalog No. AA1230730



Specifications

Specifications

Zinc chloride
7646-85-7
Cl2Zn
zinc butter
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
ACS Reagent
Hygroscopic
290°C
For analysis ACS
97%
MFCD00011295
Freely soluble in water,and dilute HCl. Soluble in alcohol,glycerol,ether and acetone.
Cl[Zn]Cl
136.28
CHEBI:49976
Lumps
2.91 g/mL
732°C
250g

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc butter
CHEBI:49976
Cl[Zn]Cl
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 231-592-0

RTECSNumber : ZH1400000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents