Please login to your online account to display your discounted pricing

Zinc chloride, 99.99%, (trace metal basis), anhydrous, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC380130050

 View more versions of this product

Catalog No. AC380130050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Free Flowing Small Crystals
Purity Grade Trace Metal Basis
Formula Weight 136.29
Quantity 5g
Color White or almost white
Density 0.95
Merck Index 15, 10331
Linear Formula ZnCl2
Molecular Formula Cl2Zn
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00011295
Chemical Name or Material Zinc chloride
Assay Percent Range 99.99%
CAS 7646-85-7
Total Trace Metal Impurities 0.01% max.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material Zinc chloride
CAS 7646-85-7
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula Cl2Zn
Linear Formula ZnCl2
Formula Weight 136.29
MDL Number MFCD00011295
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents