Learn More
Zinc chloride, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196840050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
732.0°C | |
293.0°C | |
White | |
5 g | |
Cl2Zn | |
MFCD00011295 | |
15,10331 | |
Solubility in water: 432 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and ether, freely soluble in acetone | |
Cl[Zn]Cl | |
136.29 | |
CHEBI:49976 | |
≥98% | |
Glass Bottle |
Zinc chloride | |
7646-85-7 | |
Crystalline Powder, Crystals and/or Chunks | |
98% min. (Complexometry) | |
ZnCl2 | |
01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.29 | |
Extra Pure | |
.98g/mL |
Chemical Identifiers
7646-85-7 | |
136.29 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 231-592-
RUO – Research Use Only