Please login to your online account to display your discounted pricing

Zinc Chloride, ≥97%, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0219145190

 View more versions of this product

Catalog No. ICN19145190

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range ≥97%
Molecular Formula ZnCl2
Formula Weight 136.28g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range ≥97%
Molecular Formula ZnCl2
Formula Weight 136.28g/mol
Accessories

Room Temperature