Please login to your online account to display your discounted pricing

Zinc Chloride, ≥97%, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  19145180

 View more versions of this product

Catalog No. ICN19145180

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 136.28
Quantity 100g
Physical Form Crystalline
Viscosity [NULL]
Chemical Name or Material Zinc Chloride
Molecular Formula ZnCl2
Assay Percent Range ≥97%
CAS 7646-85-7
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range ≥97%
Molecular Formula ZnCl2
Formula Weight 136.28
Accessories

Room Temperature