Please login to your online account to display your discounted pricing

Zinc Chloride, ≥97%, (Crystalline/ACS Reagent Grade), MP Biomedicals

$54.22 - $54.22

Chemical Identifiers

Chemical Name or Material Zinc Chloride
CAS 7646-85-7
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range ≥97%
Molecular Formula ZnCl2
Formula Weight 136.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19145180

  SDS
mp biomedicals
19145180
100g Each for $54.22
Description & Specifications

Specifications

Formula Weight 136.28
Purity Grade ACS Reagent Grade
Quantity 100g
Physical Form Crystalline
Chemical Name or Material Zinc Chloride
Molecular Formula ZnCl2
Assay Percent Range ≥97%
CAS 7646-85-7
Recommended Storage Room Temperature
Safety and Handling

Room Temperature