Please login to your online account to display your discounted pricing

Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC370051000

 View more versions of this product

Catalog No. AC370051000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution, May Contain A Small Amount Precipitate
Formula Weight 136.29
Quantity 100mL
Color Colorless to faintly yellow or slightly blue-gray
Packaging AcroSeal™ Glass Bottle
Density 0.8400g/mL
Concentration 0.9 to 1.1M
Fieser 01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391
Merck Index 15, 10331
Water 0.2% max. (Coulometric)
Name Note 1.0M solution in diethyl ether
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
CAS 7646-85-7
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Storage Information Product can form two phases, which can recombine on cooling
Chemical Name or Material Zinc chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
May form explosi

Specifications

Chemical Name or Material Zinc chloride
Name Note 1.0M solution in diethyl ether
CAS 7646-85-7
Molecular Formula Cl2Zn
Linear Formula ZnCl2
Formula Weight 136.29
MDL Number MFCD00011295
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
May form explosi

SDS & Safety Documents