missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals
$68.41 - $192.56
Chemical Identifiers
CAS | 5263-02-5 |
---|---|
Molecular Formula | C2H6O12Zn5 |
Molecular Weight (g/mol) | 548.96 |
MDL Number | MFCD00011519 |
InChI Key | UOURRHZRLGCVDA-UHFFFAOYSA-D |
Synonym | Zinc subcarbonate; Zinc carbonate hydroxide |
IUPAC Name | pentazinc(2+) hexahydroxide dicarbonate |
SMILES | [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1459030
|
Thermo Scientific Chemicals
A1459030 |
250 g |
Each for $68.41
|
|
|||||
AAA145900B
|
Thermo Scientific Chemicals
A145900B |
1000 g |
Each for $192.56
|
|
|||||
Description
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsZinc carbonate basic is used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
5263-02-5 | |
548.96 | |
UOURRHZRLGCVDA-UHFFFAOYSA-D | |
pentazinc(2+) hexahydroxide dicarbonate |
C2H6O12Zn5 | |
MFCD00011519 | |
Zinc subcarbonate; Zinc carbonate hydroxide | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O |
Specifications
5263-02-5 | |
C2H6O12Zn5 | |
Zinc subcarbonate; Zinc carbonate hydroxide | |
UOURRHZRLGCVDA-UHFFFAOYSA-D | |
pentazinc(2+) hexahydroxide dicarbonate | |
548.97 | |
Zinc carbonate basic |
250 g | |
MFCD00011519 | |
Insoluble in water. | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O | |
548.96 | |
97% | |
Zn >58.0% |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 226-076-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only