Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc bromide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Zinc bromide
CAS 7699-45-8
Name Note Ultra dry
Formula Weight 225.19
MDL Number MFCD00011294
UN Number UN3260
Merck Index 14,10129
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3572506

 
alfa aesar™
3572506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572514

 
alfa aesar™
3572514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572522

 
alfa aesar™
3572522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.19
Packaging Ampule
Chemical Name or Material Zinc bromide
TSCA TSCA
Mesh Size −10 Mesh
Name Note Ultra dry
Physical Form Beads
Refractive Index 1.5452
Assay 99.999%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00011294
CAS 7699-45-8
UN Number UN3260
Merck Index 14,10129
Boiling Point 650°C
Melting Point 394°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 4.201g/mL
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
RTECS Number ZH1150000

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon