missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc bromide hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1296006

 View more versions of this product

Catalog No. AA1296006



Specifications

Specifications

Zinc bromide hydrate
(metals basis)
MFCD00011294
14,10129
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zinc;dibromide
10421208
Crystalline
Puratronic™
7699-45-8
Br2Zn
UN3260
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic®, zinc bromide, anhydrous 100g
[Zn+2].[Br-].[Br-]
225.188
225.19 (Anhydrous)
99.999%
5g

Chemical Identifiers

7699-45-8
225.188
VNDYJBBGRKZCSX-UHFFFAOYSA-L
10421208
[Zn+2].[Br-].[Br-]
Br2Zn
MFCD00011294
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic®, zinc bromide, anhydrous 100g
zinc;dibromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents