Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc bromide hydrate, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Zinc bromide hydrate
CAS 7699-45-8
Purity Grade Puratronic™
Formula Weight 225.19 (Anhydrous)
MDL Number MFCD00011294
UN Number UN3260
Merck Index 14,10129
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1296006

  SDS
alfa aesar™
1296006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1296014

  SDS
alfa aesar™
1296014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1296022

  SDS
alfa aesar™
1296022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.19 (Anhydrous)
UN Number UN3260
TSCA TSCA
Assay 99.999%
CAS 7699-45-8
Merck Index 14,10129
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material Zinc bromide hydrate
Purity Grade Puratronic™
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Physical Form Crystalline
MDL Number MFCD00011294
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes (metals basis)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.