missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc bromide, anhydrous, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4422330

 View more versions of this product

Catalog No. AA4422330



Description

Description

  • Packaged under Argon
Specifications

Specifications

Zinc bromide
7699-45-8
Br2Zn
UN3260
acmc-209p7h
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zinc;dibromide
10421208
Powder,Packaged under argon
4.201 g/mL
Hygroscopic
White
250g
Anhydrous
(metals basis)
MFCD00011294
14,10129
Very soluble in ethanol and soluble in ethyl ether. Soluble in water (at 20°C 4470g/L).
[Zn+2].[Br-].[Br-]
225.188
225.19
99.999%
1.5452
650°C
394°C

Chemical Identifiers

7699-45-8
225.188
VNDYJBBGRKZCSX-UHFFFAOYSA-L
10421208
[Zn+2].[Br-].[Br-]
Br2Zn
MFCD00011294
acmc-209p7h
zinc;dibromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-718-4

RTECSNumber : ZH1150000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents