missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 46880MD

 View more versions of this product

Catalog No. AA46880MD



Specifications

Specifications

Zinc bis (trifluoromethylsulfonyl)imide
168106-25-0
>110°C (230°F)
Moisture Sensitive
[(CF3SO2)2N]2Zn
Zinc triflimide; Zinc trifluoromethanesulfonimide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2]
625.651
625.65
143°C to 147°C
250mg
White
C4F12N2O8S4Zn
MFCD16621474
QEORIOGPVTWFMH-UHFFFAOYSA-N
zinc;bis(trifluoromethylsulfonyl)azanide
131849436
Solid

Chemical Identifiers

168106-25-0
625.651
QEORIOGPVTWFMH-UHFFFAOYSA-N
131849436
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2]
C4F12N2O8S4Zn
MFCD16621474
Zinc triflimide; Zinc trifluoromethanesulfonimide
zinc;bis(trifluoromethylsulfonyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents