Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc arsenide, 99%

Chemical Identifiers

Chemical Name or Material Zinc arsenide
CAS 12006-40-5
Molecular Formula Zn3As2
Formula Weight 345.95
MDL Number MFCD00049629
UN Number UN1557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1762314

 
alfa aesar™
A1762314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1762322

 
alfa aesar™
A1762322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 345.95
Chemical Name or Material Zinc arsenide
TSCA TSCA
Odor Odorless
Assay 99%
Molecular Formula Zn3As2
MDL Number MFCD00049629
CAS 12006-40-5
UN Number UN1557
Melting Point 1015°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.