missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Acetate, For Cyanide, 10% (w/v), LabChem™

$89.03 - $89.03

Chemical Identifiers

CAS 7732-18-5,5970-45-6
CAS Max % 90,10
Linear Formula Zn(C2H3O2)2·2H2O
Formula Weight 219.49
Chemical Name or Material Zinc Acetate, For Cyanide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC270802 Lab Chem
LC270802
1L Each for $89.03
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,5970-45-6
Zn(C2H3O2)2·2H2O
Zinc Acetate, For Cyanide
90,10
219.49

Specifications

219.49
5970-45-6,7732-18-5
Soluble in water
1.1g/mL
Liquid
1.1g/mL
Carbon monoxide; Carbon dioxide
Zinc Acetate, For Cyanide
Clear, colorless
10,90
Room Temperature
Passes Test
Zn(C2H3O2)2·2H2O
Poly Bottle
1L
10% (w/v)

Safety and Handling

GHS H Statement
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Warning