Please login to your online account to display your discounted pricing

Zinc Acetate, Dihydrate, ACS Grade, 98.0% to 101.0%, LabChem™

$106.43 - $106.43

Chemical Identifiers

Chemical Name or Material Zinc Acetate
CAS 5970-45-6
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula Zn(C2H3O2)2·2H2O
Formula Weight 219.49
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC270701

 
lab chem
LC270701
500g Each for $106.43
Description & Specifications

Specifications

Assay 98.0 to 101.0%
Formula Weight 219.49
Chemical Name or Material Zinc Acetate
Identification Passes Test
Name Note Dihydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 1735kg/m3
Storage Information Room Temperature
Linear Formula Zn(C2H3O2)2·2H2O
CAS 5970-45-6
UN Number UN3077
Decomposition Information Carbon dioxide; Carbon monoxide
Color White
Melting Point 237°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Density 1,735kg/m3
Safety and Handling

Warning

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Very toxic to aquatic life.