missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc, 99.999% (trace metal basis), powder, -40 mesh, ACROS Organics™

$196.30 - $196.30

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym blue powder
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367260100 SDS View Documents Acros Organics
AC367260100
10g Glass bottle Each for $196.30
 
AC367260500 SDS View Documents Acros Organics
AC367260500
50g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

for ultra trace analysis, Reduction reagent
Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
blue powder
CHEBI:30185
[Zn]

Specifications

10g
7.14
Zn
MFCD00011291
15, 10326
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
Glass bottle
CHEBI:30185
Powder
Trace Metal Basis
Gray
907.0°C
7440-66-6
100.00%
Zn
01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
blue powder
[Zn]
65.38
23994
65.38
99.999%
0.001% max.
420.0°C
Zinc, 99.999%
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
Catches fire spontaneously if exposed to air.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger