Please login to your online account to display your discounted pricing

Zinc, 99.995%, (trace metal basis), powder, ACROS Organics™

$704.59 - $704.59

Chemical Identifiers

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade For trace metal basis analysis
Physical Form Powder
Assay Percent Range 100.00%
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.36
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194500500

  SDS
thermo fisher scientific
AC194500500
Glass Bottle in Tin Can 50g Each for $704.59

AC194500050

  SDS
thermo fisher scientific
194500050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 65.36
Quantity 50g
Packaging Glass Bottle in Tin Can
Purity Grade For trace metal basis analysis
Physical Form Powder
Density 7.14g/mL
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
CAS 7440-66-6
Linear Formula Zn
Assay Percent Range 100.00%
Molecular Formula Zn
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
Catches fire spontaneously if exposed to air.
Chemical Name or Material Zinc
MDL Number MFCD00011291
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

for ultra trace analysis, Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
Catches fire spontaneously if exposed to air.