Please login to your online account to display your discounted pricing

Zinc, 99.8% min., ACS, EMD Millipore


Secondary Reference Material For Complexometry

Quantity 500g

Manufacturer: emd millipore corporation  ZX00101

 View more versions of this product

Catalog No. MZX00101

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Granular Solid
Formula Weight 65.39
Color Silver metallic
Quantity 500g
Density 7.13g/cm3 (20°C)
Purity Grade SRM Certipure™
Chemical Name or Material Zinc
Name Note Traceable to NIST
Assay Percent Range 99.8% min.
Molecular Formula Zn
Linear Formula Zn
CAS 7440-66-6
Recommended Storage Store at +15°C to +25°C

Specifications

Chemical Name or Material Zinc
Name Note Traceable to NIST
CAS 7440-66-6
Purity Grade SRM Certipure™
Physical Form Granular Solid
Assay Percent Range 99.8% min.
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.39
Accessories

Store at +15°C to +25°C