Please login to your online account to display your discounted pricing

Zinc, 99.8% min., ACS, EMD Millipore

Secondary Reference Material For Complexometry

$382.71 - $3,452.71

Chemical Identifiers

Chemical Name or Material Zinc
Name Note Traceable to NIST
CAS 7440-66-6
Purity Grade SRM Certipure™
Physical Form Granular Solid
Assay Percent Range 99.8% min.
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.39
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MZX00101

 
emd millipore corporation
ZX00101
500g Each for $382.71

MZX00107

 
emd millipore corporation
ZX00107
12kg Each for $3,452.71
Description & Specifications

Specifications

Color Silver metallic
Physical Form Granular Solid
Formula Weight 65.39
Quantity 500g
Density 7.13g/cm3 (20°C)
Purity Grade SRM Certipure™
CAS 7440-66-6
Linear Formula Zn
Molecular Formula Zn
Assay Percent Range 99.8% min.
Chemical Name or Material Zinc
Name Note Traceable to NIST
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C