Please login to your online account to display your discounted pricing

Zinc, 98+%, dust (stable acc. to UN classification class 4), ACROS Organics™

$64.33 - $269.04

Chemical Identifiers

Chemical Name or Material Zinc
CAS 7440-66-6
CAS 1314-13-2
Assay ≥98.0% (Metallic Zinc)
Assay Percent Range 98+%
Molecular Formula Zn
Linear Formula Zn
Formula Weight 65.38
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC198340100

  SDS
thermo fisher scientific
AC198340100
Plastic bottle 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC198340010

  SDS
thermo fisher scientific
198340010
Plastic bottle 1kg Each for $117.92

AC198340025

  SDS
thermo fisher scientific
198340025
Plastic bottle 2.5kg Each for $269.04

AC198341000

  SDS
thermo fisher scientific
AC198341000
Plastic bottle 100g Each for $64.33

AC198340250

  SDS
thermo fisher scientific
AC198340250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0% (Metallic Zinc)
Formula Weight 65.38
Physical Form Very Fine Powder
Color Gray
Quantity 10kg
Density 7.1400g/mL
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Packaging Plastic bottle
Assay Percent Range 98+%
Linear Formula Zn
CAS 1314-13-2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Zinc
MDL Number MFCD00011291
Molecular Formula Zn
CAS 7440-66-6

Reduction reagent

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.